Learn More
Thermo Scientific™ Zirconium Oxide Round Bottom Crucibles, Low Form
Click to view available options
Capacity (Metric):
5 mL
100 mL
200 mL
Diameter (Metric) Outer:
29mm
67mm
82mm
Height (Metric):
21mm
51mm
63mm
Unit Size:
Each
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Material | Zirconium Oxide |
| Quantity | 1 Piece |
| Capacity (Metric) | 100 mL |
| Diameter (Metric) Outer | 67mm |
| Height (Metric) | 51mm |
| Product Type | Zirconium Oxide Round Bottom Crucibles, Low Form |
| Diameter (Metric) Base | 35mm |