Learn More
Thermo Scientific™ Zirconium Crucible
Click to view available options
Diameter (Metric) Outer:
39 mm
42 mm
47 mm
Height (Metric):
33 mm
35 mm
36 mm
44 mm
Description:
Flanged Rim OD: 44 mm
Flanged Rim OD: 48 mm
Flanged Rim OD: 54 mm
SlideOn Ring OD: 48 mm
SlideOn Ring OD: 54 mm
SnapOn Ring OD: 44 mm
SnapOn Ring OD: 48 mm
Snapon Ring OD: 50mm
Unit Size:
Each
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Description | Flanged Rim OD: 44 mm |
| Material | Zirconium |
| Diameter (Metric) Outer | 39 mm |
| Height (Metric) | 33 mm |
| Product Type | Zirconium Crucible |
| Diameter (Metric) Base | 29mm |