Learn More
Hach™ ZincoVer™ 5 Zinc Reagent Powder Pillows
Ready-to-use, premeasured powder reagent in individual sealed packages for determination of zinc by the ZincoVer™ 5 zincon method. US EPA approved for wastewater analysis.
Brand: Hach™ 1403268
This item is not returnable.
View return policy
Description
Powder Pillows make titration applications quick and easy.
- Sealed packets ensure fresh, uncontaminated reagent every time
- Premeasured to avoid measurement error and contamination from handling
- Ready to use packets save time and hassle
- Exact doses to easily monitor supply and prepare for lab procedures
- Single-use packs for easy transportation and packing in test kits
Specifications
Zinc | |
Individual Packet | |
25 | |
0.01 to 2.00mg/L Zn | |
Zincon |
US EPA | |
Zinc determination | |
25 Pack | |
50 mL | |
Reagent packet |