Learn More
Invitrogen™ xMAP Sheath Fluid Plus
xMAP Sheath Fluid Plus is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments
Brand:  Invitrogen™ 4050021
72.91 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Product Code. 17183935
  • € 78.25 / Each
Edge
Explore more special offers
Description
Description
  • xMAP Sheath Fluid Plus is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments. It is needed to operate all Luminex 200 and FLEXMAP 3D systems.
  • This reformulated sheath product replaces the original xMAP Sheath Fluid (Cat. No. 4050015).
  • The new sheath formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.
For Research Use Only. Not for use in diagnostic procedures. Not intended for human or animal therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 L
Container
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB97707 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number4050021
Lot NumberC01190 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number4050021
Lot NumberB96436 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number4050021
Lot NumberC09353 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number4050021
Lot NumberB95314 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number4050021
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ xMAP Sheath Fluid Plus > 20 L