Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-XT Syringe for LC
Product Code. 15239273
Click to view available options
Volume (Metric):
100 μL
250 μL
500 μL
1 mL
Diameter (Metric) Inner:
0.17 mm
0.375 mm
Unit Size:
Each
Product Code. 15239273
Brand: Trajan Scientific and Medical™ 2928630
The knowledge and expertise that goes into manufacturing an SGE syringe gives outstanding performance and superior quality
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
Specifications
Description CTC PAL-XT Syringe for LC
Needle Gauge 22 G
Diameter (Metric) Inner Needle 0.375 mm
Diameter (Metric) Inner 0.375 mm
Diameter (Metric) Outer 0.71 mm
Diameter (Metric) Outer Needle 0.71 mm
For Use With (Application) For liquid handling and LC
Volume (Metric) 250 μL
Needle Point Style LC