Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-XT Syringe for GC
Click to view available options
Volume (Metric):
100 μL
250 μL
1 mL
Diameter (Metric) Inner:
0.15 mm
0.32 mm
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
Specifications
Specifications
| Description | CTC PAL-XT Syringe |
| Needle Gauge | 23 G |
| Diameter (Metric) Inner Needle | 0.32 mm |
| Diameter (Metric) Inner | 0.32 mm |
| Diameter (Metric) Outer | 0.63 mm |
| Diameter (Metric) Outer Needle | 0.63 mm |
| For Use With (Application) | For gas chromatography, liquid handling |
| Volume (Metric) | 1 mL |
| Needle Point Style | Bevel |