Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-xt Autosampler Syringes
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
Diameter (Metric) Outer Needle:
0.63 mm
0.71 mm
Material:
Metal (Plunger)
PTFE (Plunger)
PTFE-tipped (Plunger)
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Available as all-metal or PTFE-tipped plunger
Specifications
Specifications
| Material | Metal (Plunger) |
| Description | Autosampler Syringe |
| Needle Gauge | 22 G |
| Diameter (Metric) Inner Needle | 0.17 mm |
| Diameter (Metric) Outer Needle | 0.71 mm |
| Volume (Metric) | 10 μL |
| Needle Point Style | LC |