Learn More
Waters Corporation XBridge Prep C18 Column
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12).
Brand:  Waters Corporation 186003898
Encompass
 View more versions of this product
Product Code. 13213250
  • € 12030.00 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Description
  • The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
Specifications
Specifications
50 mm
6000 psi (415 Bar)
10 μm
1/Ea.
Yes
50 mm
Spherical
130 Å
L1
Column