missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Sterilin X700 container aseptice 7ml no label
GREENER_CHOICE

Brand:  Thermo Scientific Sterilin M00525

161.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


X700 container aseptice 7ml no label

Product Code. 12799985

  • € 270.00 / Pack of 700

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Sterilin X700 container aseptice 7ml no label >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.