missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nutrient Agar (Slant)

Product Code. 11985345
Click to view available options
Fill Weight:
7 to 8 g
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11985345

Brand: Thermo Scientific™ TV5011Z

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Cultivate and maintain a wide variety of non-fastidious bacteria for routine culture, isolation, and storage.

Nutrient Agar (Slant) is used to cultivate and maintain a wide variety of non-fastidious bacteria for routine culture, isolation, and storage.

TRUSTED_SUSTAINABILITY

Specifications

Color Sand Yellow, Transparent
Content And Storage 2°C to 25°C
Format Poured Tube, Slant Agar
Manufacturing Origin Germany
Product Type Agar
Fill Weight 7 to 8 g
Packaging Type 50 Tubes/Box
Quantity 50/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.