Learn More
Fisherbrand™ Connecting Cable
Connecting cable with BNC Plug for combined ion-sensitive electrode with solid membranes that is ideal for measuring agriculture, water analysis, and calibration of ionic electrodes.
Brand: Fisherbrand™ 90603
This item is not returnable.
View return policy
Description
- Connecting Cable with BNC Plug
- For combined ion-sensitive electrode with solid membranes
- For use with measuring agriculture, water analysis, and calibration of ionic electrodes
Specifications
BNC | |
Connecting Cable | |
For connecting the electrode to a pH meter, using an anti -static coaxial cable with the correct plug . | |
3 m |