Learn More
Thermo Scientific™ Baird Parker RPF Agar Base
Click to view available options
Fill Weight:
89 to 91 g
Unit Size:
Pack of 10
Description
Baird Parker RPF Agar Base is a ready to use selective medium to isolate and enumerate coagulase‐positive staphylococci from food samples.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 25°C |
| Format | 125 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 89 to 91 g |
| Packaging Type | 10 Sirop Bottles with metal R.O.P.P. cap closures in a box |
| Quantity | 10/Pk. |