missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ Bis-Tris Midi Protein Gels, 10%, 1.0 mm, WedgeWell™ format

Product Code. 30132171
Click to view available options
Wells:
12 + 2-well
20-well
26-well
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 30132171

Brand: Invitrogen™ WBT01012BOX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Invitrogen NuPAGE Bis-Tris protein gels, WedgeWell format are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial.

Invitrogen NuPAGE Bis-Tris protein gels, WedgeWell format are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Invitrogen NuPAGE Bis-Tris WedgeWell format protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Product features

  • Better protein integrity—optimized sample preparation process preserves your proteins
  • Faster run times—get separation of your proteins in as little as 30 minutes
  • Longer shelf life—NuPAGE Bis-Tris gels can be stored for 12 months at room temperature
  • Wedge-shaped wells—for easier sample loading and up to two times higher sample volume

Choose the right NuPAGE Bis-Tris gel for your protein separation

NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Sample Reducing Agent (NP0004). NuPAGE Antioxidant (NP0005) can be added to the running buffer to maintain the reduced state of the proteins during the run allowing for maximum band sharpness if desired. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins with a faster run time or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.

NuPAGE WedgeWell Bis-Tris Midi gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100).

For transfer of proteins to a membrane, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001). Alternatively, traditional wet transfer can be performed using the SureLock Tandem Midi Blot Module (STM2001) using NuPAGE Transfer buffer (NP00061).

Convenient to use

NuPAGE Bis-Tris WedgeWell format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4–25°C. Do not freeze.
For Use With (Equipment) XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank
Gel Percentage 10%
Gel Size Midi
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Sample Loading Volume Up to 100 μL
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 12 + 2-well
Mode of Separation Molecular Weight
Product Line NuPAGE
Quantity 10 gels (1 box)
Shipping Condition Room Temperature
Length (Metric) 8 cm
Width (Metric) 13 cm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.