Learn More
Corning™ X-SERIES™ Counterweight
Shop All Corning Products
Click to view available options
Product Type:
Weight: High
Weight: Low
Weight: Medium
Capacity (Metric):
161 to 240 mL
40 to 90 mL
91 to 160 mL
Unit Size:
Each
Description
The Corning™ X-SERIES™ Systems are manufactured for Corning™ by ThermoGenesis Corp.
X-SERIES™, X-WASH™, X-LAB™, X-BACS™, and X-MINI™ are either registered trademarks or trademarks of ThermoGenesis Corp. in the United States and/or other countries.
Specifications
Specifications
| Capacity (Metric) | 40 to 90 mL |
| For Use With (Equipment) | X-WASH and X-LAB Systems |
| Quantity | 1/Pk., 1/Cs. |
| Product Type | Weight: Low |