Learn More
Draeger Safety™ X-Plore 6300 Full Face Mask
The X-Plore 6300 Full Face Mask is for the price conscious user who demands quality and wearing comfort. The double layered face seal for triple sealing action ensures a comfortable and tight fit. The visor is made of highly scratch resistant plexiglass (PMMA). Without neck strap.
Brand: Draeger Safety™ R55800
This item is not returnable.
View return policy
Description
- Secure and comfortable fit
- Mask body made from durable and skin-friendly EPDM
- Double-layer face seal and triple sealing edges for a secure and comfortable seal for every face type
- 180° panoramic view
- Scratch-proof and distortion-free PMMA plexiglas visor
- Efficient ventilation system - separate inner mask prevents the lens from fogging
- 5-point harness - donned easily and quickly
- Easy maintenance and logistics
- Universal fit
- Barcode on the inside simplifies recording and maintaining of the mask inventory
- Versatile use
- Equipped with a Rd40 connection in accordance with EN 148-1 and can be combined with Rd40 filter range
Specifications
CE, EN 136 | |
Standard thread | |
Full Face Mask | |
Personal | |
Anti-fog, Anti-scratch | |
X-plore™ 6300 | |
1 Ea. |
EN 136 Class 2 (CE mark), NIOSH, AS/NZS | |
Black | |
Yes | |
Adjustable 5-point | |
Polycarbonate | |
No |
Product Content Correction
Product Title
Draeger Safety™ X-Plore 6300 Full Face Mask > X-Plore 6300 Full Face Mask; Lens Material: Polycarbonate