missing translation for 'onlineSavingsMsg'
Learn More

Endecotts™ Woven Wire Mesh Sieve

Available in a variety of diameters and apertures

Brand:  Endecotts™ 200SIW.125

 View more versions of this product

Product Code. 10173924

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Endecotts Woven Wire Mesh Sieves are made of stainless steel and available in a wide variety of diameters and apertures.

  • Woven Wire Mesh Sieve
  • Made of Stainless Steel
Specifications

Specifications

125 μm
7.87 in.
200 mm
Woven Mesh
Sieve
Stainless Steel
Stainless Steel
8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products

Accessories and Services

Optional Accessory

Show More Show Less

Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Endecotts™ Woven Wire Mesh Sieve > Diameter: 200 mm, Pore Size: 125 μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.