Learn More
BD Plastipak™ Wound and Bladder Syringe
Eliminates the risk of allergic reactions to latex. BD™ Plastipak™ Wound and Bladder Syringe features a latex-free stopper with a slight and uniform sliding action. The clear barrel with boldly printed 1mL graduations enables perfect visualization of the syringe contents and volume.
Brand:  BD 300867
Product Code. 10440864
  • € 265.00 / Pack of 60
Edge
Explore more special offers
Description
Description
  • Very high tightness due to tight, double-sealing ring
  • Retaining ring prevents accidental plunger rod withdrawal
  • Centered catheter tip attachment
  • Silicone oil lubricant ensures piston slides gradually and evenly
  • Spray cylinder and piston rod made of polypropylene (PVC-free)
  • Disposable syringes are EO-sterilized and individually packaged
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
29.4 mm
133 mm
Single-use
60 Pk.
Centered Catheter
Clear
Yes
Wound/Bladder Syringe
1 mL
50 mL
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers