Learn More
Woodsafe™ Woodsafe Sharpsbin Holders
Product Code. 18742815
Click to view available options
For Use With (Equipment):
0.5 L Woodsafe Containers
2 and 3 L Woodsafe Containers
25 L and 50 L Woodsafe Containers
6 L and 12 L Woodsafe Containers
Woodsafe Holders
Product Type:
Holder
String Hook
Table Mount
Trolley
Unit Size:
Each
Pack of 10
Pack of 100
Product Code. 18742815
Brand: Woodsafe™ 5120028
Only null left
Add to basket
Add to basket
Accessories to Woodsafe sharpsbins and clinical waste bins. Holders and trolleys.
  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Steady holders and trolleys
Specifications
Description Woodsafe Sharpsbin Holders
For Use With (Equipment) 6 L and 12 L Woodsafe Containers
Product Type Holder
Lower climate impact and ACT label