Learn More
Reichert Instruments™ Wolffheugel Counting Plate
The Darkfield Quebec colony counter allows the user to quickly and accurately count bacteria colonies
Brand:  Reichert Instruments™ 13333800
Product Code. 12762585
  • € 208.00 / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Description
The Darkfield Quebec colony counter allows the user to quickly and accurately count bacteria colonies.
It is available in two models for digital or manual counting. Both models provide even, glare-free illumination as light is spread uniformly over the entire culture plate.
Adjustable dish holder for centering both round dishes and square culture plates
  • Adjustable focusing rod
  • Lens rotates a full 360°
  • Built-in tilt leg
  • Optional 1.5x auxiliary lens fits over standard lens, increasing magnification to 3x
Specifications
Specifications
Counting Plate
1Each
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Reichert Instruments™ Wolffheugel Counting Plate >