Learn More
Bode Chemie™ Wipes
€ 92.00
Specifications
Product Type Wipes
For Use With (Application) Cleaning disinfection of alcohol-sensitive surfaces
For Use With (Equipment) sensitive medical devices
View More Specs
Products 1
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
10433475
View Documents
Bode Chemie™
9813875
180 x 200 mm
€ 92.00
80 sheets
In Stock
to see stock.
Specifications
Wipes
Cleaning disinfection of alcohol-sensitive surfaces
sensitive medical devices
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Bode Chemie™ Wipes