Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Screw-Cap NMR Tube
Product Code. 15318998 Shop All SP Scienceware Products
Click to view available options
Length (Metric):
177.8 mm
228.6 mm
Model:
507-TR-7
513-7TRA-9
528-TR-7
Field Strength:
300 MHz
400 MHz
500 MHz
Unit Size:
Each
Pack of 5
Product Code. 15318998
Brand: Bel-Art™ SP Scienceware™ 665051095
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Screw-Cap NMR Tube is commonly used in sample degasification due to its capability to maintain vacuum quality.
  • Each Screw-Cap Tube comes complete with open top screw cap and PTFE/Silicone septum
  • Inert to most organic solvents and compounds but poor to strongly corrosive materials
  • Remains reliable after multiple punctures
Specifications
Length (Metric) 228.6 mm
Material Borosilicate Glass
Product Type High Precision NMR Sample Tube
Diameter (Metric) Outer 5 mm
Field Strength 400 MHz
Includes PTFE/Silicon Septum
Model 513-7TRA-9