Learn More
Sampling Systems™ Wide Sampler Cleaning Brushes
Product Code. 15809411 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
600 mm
1100 mm
1600 mm
2100 mm
Unit Size:
Each
Product Code. 15809411
Brand: Sampling Systems™ 8020J1600
Only null left
Add to basket
Add to basket
Ensure that the sampler is properly cleaned in order to prevent cross contamination. Sampling Systems™ Wide Sampler Cleaning Brushes are a range of brushes that are ideal for cleaning both stainless steel and plastic samplers.
  • Hygienic nylon brushes
  • Robust aluminum handle
  • Wide brush shape
Specifications
Product Type Cleaning Brush
Handle Material Aluminum
Diameter (Metric) Bristle 45 mm
For Use With (Equipment) Tubes with 30 to 45mm Diameter
Length (Metric) 1600 mm