Learn More
Gerresheimer™ Threaded Clear Wide Mouth Bottles without Closure
Stock the laboratory with Gerresheimer™ Threaded Clear Wide Mouth Bottles without Closure. The round, glass bottles are available in various capacities.
Brand: Gerresheimer™ 4456063002
Description
- Round
- Wide Mouth
- Clear Glass
- Without closure
Specifications
30 mL | |
Glass | |
Wide |
Clear | |
126Pack | |
Round Bottle |
Product Content Correction
Product Title
Gerresheimer™ Threaded Clear Wide Mouth Bottles without Closure > Capacity: 30mL