Learn More
IKA™ WiCo RET Control-visc
Wireless Controller (WiCo) is used for wireless remote controlling of LR 1000 control and RET control-visc.
Brand:  IKA™ 0020007259
Product Code. 16268451
  • € 494.00 / Each
Explore more special offers
Description
Description
  • The WiCo is equipped with a USB socket (Universal Serial Bus) with which the WiCo can be connected with station, the USB icon appears on the screen
  • If the WiCo is not connected with station via a USB cable, the data exchange between the station and the WiCo via BluetoothR
  • In this case, the BluetoothR icon is displayed
  • Depending on the structure of the building, the WiCo can be operated at a distance up to 15 m from the station, using the BluetoothR connection
Specifications
Specifications
5 to 40°C
Magnetic Stirrer Remote
0.3 kg
105 x 160 x 40 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers