missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ White PTFE Tubing
White PTFE Tubing is temperature resistant up to 270°C in the absence of active pressure.
Brand: Deutsch Neumann™ 3400813
This item is not returnable.
View return policy
Description
- Manufactured from polytetrafluorethylene (PTFE)
- Minimum order length of 5 m
- Almost totally chemically inert
- Excellent flexibility and strength, even in low temperatures
Specifications
ISO 9001:2008 | |
0.8 mm | |
1.3 mm | |
0.25 mm | |
White |
Plastic Tubing | |
PTFE | |
5 m | |
1 Ea. | |
270°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction