missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ White PTFE Tubing

White PTFE Tubing is temperature resistant up to 270°C in the absence of active pressure.

Brand:  Deutsch Neumann™ 3400406

 View more versions of this product

Product Code. 10036401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Manufactured from polytetrafluorethylene (PTFE)
  • Minimum order length of 5 m
  • Almost totally chemically inert
  • Excellent flexibility and strength, even in low temperatures
Specifications

Specifications

ISO 9001:2008
PTFE
1 mm
White
4 mm
6 mm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Deutsch Neumann™ White PTFE Tubing > Inner Diameter: 4 mm; Outer Diameter: 6 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.