Learn More
Mutexil™ Non-woven White Protective Clip Caps
Click to view available options
Size:
19 in.
20 in.
Description
- Polypropylene
- Disposable
- Bouffant style
- Pleated
- White
Specifications
Specifications
Product Type | Peaked Bouffant Cap |
Size | 20 in. |
Color | White |
Disposable | Single-use |
Quantity | 1000/Pk. |