Learn More
Fisherbrand™ White Plastic General Purpose Trays
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
460 x 260mm
570 x 340mm
570 x 540mm
680 x 540mm
700 x 460mm
1130 x 540mm
Unit Size:
Each
Description
- Available in general purpose, biohazard and radiation safety formats
- Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
- All trays supplied with one free liner when purchased
- Additional environmentally friendly APET liners are provided in packs of 25
Specifications
Specifications
| Dimensions (L x W) | 680 x 540mm |
| Color | White |
| Length (Metric) | 680 mm |
| Material | Plastic |
| Type | Tray |
| Width (Metric) | 540 mm |
| For Use With | Safe containment of spillages, lab bench protection |
| Includes | Liner |
| No. per Pack | 1 Each |
| Shape | Rectangular |