Learn More
Moldex™ White Face Mask
Extremely low breathing resistance increases the performance of both mask and wearer
Brand: Moldex™ 320501
Description
The uniquely developed Series 3000 Air from Moldex offers extremely low breathing resistance and therefore increased performance of both mask and wearer. The filtering surface is almost three times larger than a standard filter product, allowing you to breathe more easily.Specifications
EN 149:2001A1:2009 | |
Single-use | |
Face Mask | |
Protects against dust, mist and fumes | |
30 g |
White | |
Filter Layer, Inner Shell, DuraMesh: Polypropylene, Ethylene-vinyl acetate (EVA), Nose Seal, Clip: Polyethylene, Head Strap: Polyester, Lycra, Ventex-valve: Natural Rubber | |
Medium/Large | |
10/Box |