Learn More
DWK Life Sciences Wheaton™ V Vial™ with Solid-Top Screw Cap, Clear
Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
0.3 mL
1 mL
2 mL
2.0 mL
3 mL
5 mL
5.0 mL
10 mL
Height (Metric):
35 mm
44 mm
50 mm
61 mm
65 mm
72 mm
Graduated:
No
Yes
Closure Size:
13-425
15-415
20-400
Unit Size:
Pack of 12
Pack of 6
Description
Includes
14B rubber-lined black phenolic screw caps faced with PTFE
Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
Specifications
Specifications
| Material (Liner) | Linerless |
| Color | Clear |
| Diameter (Metric) Outer | 17 mm |
| Material | Borosilicate Glass (Low Extractable) |
| Volume (Metric) | 2.0 mL |
| For Use With (Application) | Ideal for small scale reactions, centrifugation, storage, packaging and shipping |
| Graduated | Yes |
| Style | Conical V |
| Thread Style | Screw |
| Volume (English) | 0.5 dr. |
| Show More |