Learn More
DWK Life Sciences Wheaton™ Safety-Coated Reagent Bottles
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
250 mL
Description
Features convenient writing patch.
- Clear borosilicate glass
- Attached 33-430 screw cap
- Polypropylene cap with PTFE liner
- Autoclavable
Specifications
Specifications
Capacity | 8.4 oz., 250mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8.4 oz. |
Color | Clear |
Material | Glass |
Closure Color | White |
Certifications/Compliance | ISO 9001 |
Closure Included | Yes |
Height (Metric) | 151 mm |
Closure Liner | Polyethylene Foam |