Learn More
DWK Life Sciences Wheaton™ Reusable PVC Coliwasas and Accessories
Product Code. p-322937 Shop All Wheaton Science Products Products
Click to view available options
Product Type:
Coliwasa Sampler
Extension and Connector
Length (Metric):
1.21 m
1.82 m
Length (English):
4 ft.
6 ft.
Unit Size:
Each
Product Code. 11734129
Brand: DWK Life Sciences 885253
Only null left
Add to basket
Add to basket
For a variety of applications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Extension and Connector
Capacity (English) 25.6 oz.
Capacity (Metric) 760 mL
For Use With (Equipment) Clear PVC reusable coliwasas
Length (English) 4 ft.
Length (Metric) 1.21 m