missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Orange Product Code.: 12456111
Add to basket

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Orange

Product Code. 12456111 Shop All Wheaton Science Products Products
15-415, Plastic dropping bottle, 1000 Cs.
Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
No. per Case:
100 Cs.
1000 Cs.
This item is not returnable. View return policy

Product Code. 12456111

Brand: DWK Life Sciences W242513A

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Material Polypropylene
No. per Case 1000 Cs.
Neck Finish 15-415
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Plastic dropping bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Orange >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.