Learn More
DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
3000 mL
6 L
Includes:
Flask only
Flask with impeller
Unit Size:
Each
Description
- Borosilicate glass flasks
- Two large side arms with screw-cap closures
- Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
- Graduated
- Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
- Autoclavable flasks and impellers for selected sizes can also be ordered separately
- U.S. Patent No. 3,854,704
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Material | Borosilicate Glass |
| Includes | Flask only |
| Side Arm Joint | 33-430 |
| Autoclavable | Autoclavable |
| Graduated | Yes |
| Closure Size | 51-440 |