Learn More
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips
Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
Unit Size:
Pack of 100
Pack of 1000
Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL
Specifications
Specifications
| Color | Gray |
| Material | LDPE |
| No. per Case | 100 Cs. |