missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips

Product Code. 10285702 Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
Unit Size:
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 10285702

Brand: DWK Life Sciences W24240101A

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

8mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Diameter (Metric) Inner 8 mm
Material LDPE
No. per Case 1000 Cs.
Certifications/Compliance ISO 9001
Drop Size 41 μL
Shape Round
Product Type Dropper Tip
For Use With (Equipment) Plastic dropping bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.