Learn More
Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)
13mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
Brand:  Wheaton™ W242411-01-A
Product Code. 15162419
  • ONLINE EXCLUSIVE PRICE
    € 331.00
    € 327.57 / Pack of 1000
    Save € 3.43
    1% Off
Only null left
Estimated Shipment: 25-03-2026
Description
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000