missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Dounce Tissue Grinders

Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.

Brand:  DWK Life Sciences 357544

 View more versions of this product

Product Code. 10374171

This item is not returnable. View return policy

Description

Description

Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.

  • Sample covers the pestle ball, preventing heat buildup by reducing friction against the mortar
  • Mortar features large reservoir and pouring lip
  • Supplied with “loose” and “tight” pestles
  • Components are made of low extractable borosilicate glass

 

Order Info

  • Tight pestle clearance: 0.0020 ±0.001 in. (0.05 ±0.025mm)
  • Loose pestle clearance: 0.114 ±0.001 in. (0.114 ±0.025mm)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5 oz.
Borosilicate Glass
Designed to retain a high percentage of cell nuclei, mitochondria and Proteins in soft tissues or cell suspensions
210mm
94mm
22mm
15 mL
Dounce Tissue Grinder
8.26 in.
3.7 in.
0.86 in.
0.5 oz., 15mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Dounce Tissue Grinders > 15mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.