Learn More
DWK Life Sciences Wheaton™ Decappers
Product Code. p-206810 Shop All Wheaton Science Products Products
Click to view available options
Closure Type:
13 mm Caps
30 mm Caps
Type:
Hand-operated Decapper
Plier-style Decapper
Unit Size:
Each
Product Code. 10628332
Brand: DWK Life Sciences 224357
Only null left
Add to basket
Add to basket
These crimpers are used for flip cap seals and for aluminum seals
TRUSTED_SUSTAINABILITY
Specifications
Closure Type 30 mm Caps
Type Hand-operated Decapper
For Use With (Equipment) Flip Cap Seals and Aluminum Seals