Learn More
DWK Life Sciences Wheaton™ Amber Glass E-Z Vials Convenience Packs
Product Code. 12484911 Shop All Wheaton Science Products Products
Click to view available options
Color (Seal):
Blue
Green
Natural
Red
Material (Liner):
PTFE/Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Unit Size:
Pack of 100
Product Code. 12484911
Brand: DWK Life Sciences W2251730203
Only null left
Add to basket
Add to basket
Wheaton E-Z Vials have a 40% larger opening improves sample accessibility while reducing autosampler needle damage.
  • Manufactured from certified clear and amber Type I borosilicate glass
  • Vials are packaged 100 per shelf pack and 10 packs per case
  • 40% larger opening improves sample accessibility while reducing autosampler needle damage
TRUSTED_SUSTAINABILITY
Specifications
Color (Seal) Red
Material (Liner) PTFE/Silicone
Color Amber
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 1.8 mL
Septum 0.005 PTFE/0.05 Silicone r/w Liner
Height (English) 1.25 in.
Material (Seal) Aluminum
Neck Style Crimp Top
Closure Type Crimp Cap
Height (Metric) 32 mm
Quantity 100 Pk.
Closure Size 11 mm
Marking Spot Yes