Learn More
DWK Life Sciences Wheaton™ Amber Glass E-Z Vials Convenience Packs
Product Code. 12444911 Shop All Wheaton Science Products Products
Click to view available options
Color (Seal):
Blue
Green
Natural
Red
Material (Liner):
PTFE/Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Unit Size:
Pack of 100
Product Code. 12444911
Brand: DWK Life Sciences W2251730103
Only null left
Add to basket
Add to basket
Wheaton E-Z Vials have a 40% larger opening improves sample accessibility while reducing autosampler needle damage.
  • Manufactured from certified clear and amber Type I borosilicate glass
  • Vials are packaged 100 per shelf pack and 10 packs per case
  • 40% larger opening improves sample accessibility while reducing autosampler needle damage
TRUSTED_SUSTAINABILITY
Specifications
Color (Seal) Red
Material (Liner) PTFE/Rubber
Color Amber
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 1.8 mL
Septum 0.002 PTFE/0.037 Rubber Liner
Height (English) 1.25 in.
Material (Seal) Aluminum
Neck Style Crimp Top
Closure Type Crimp Cap
Height (Metric) 32 mm
Quantity 100 Pk.
Closure Size 11 mm
Marking Spot Yes