Learn More
DWK Life Sciences Wheaton™ ABC Clear Glass Vial Convenience Packs
Product Code. 12474811 Shop All Wheaton Science Products Products
Click to view available options
Marking Spot:
No
Closure Color:
Black
Blue
Green
Natural
Yellow
Septum:
Blue PTFE, White Silicone
Red PTFE
Red PTFE, White Silicone
Red PTFE, White Silicone, Red PTFE
Pre-slit:
No
Yes
Unit Size:
Pack of 100
Product Code. 12474811
Brand: DWK Life Sciences W2251500201
Only null left
Add to basket
Add to basket
40% larger opening improves sample accessibility while reducing autosampler needle damage
  • Manufactured from clear Type I borosilicate glass
  • 12 x 32 mm screw top with 9 mm ABC screw cap
  • Available with and without writing patch
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Diameter (English) Outer 0.47 in.
Color Clear
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Pre-slit No
Volume (Metric) 1.8 mL
Septum Red PTFE, White Silicone
Height (English) 1.25 in.
Closure Material Polypropylene
Closure Type Screw Cap
Height (Metric) 32 mm
Closure Size 9 mm
Marking Spot No