missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences 520 Bottle-Top Dispenser Reservoirs

Large selection of glass and Polyethylene reagent bottles, each supplied with one screw cap. Perfectly adapted with Calibrex™ bottle top dispensers.

Brand:  DWK Life Sciences 844025

27.12 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12427191

  • € 30.15 / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Corresponding neck adapters supplied with the dispensers.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Reagent Bottle
Soda Lime Glass
520 model bottletop dispenser
8.45 oz.
32 mm
Amber
250 mL
Square
1.25 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers