Learn More
Cytiva Whatman™ Specialized Test Paper
Product Code. 11944426
Click to view available options
For Use With (Application):
pH
Unit Size:
Pack of 10
Product Code. 11944426
Brand: Cytiva 2651500
Only null left
Add to basket
Add to basket
Book, starch iodide.
  • Instant pH readings.
  • Accurate for a wide range of routine pH testing.
  • Inexpensive.
  • Convenient and portable for field use.
TRUSTED_SUSTAINABILITY
Specifications
Color Indicator Starch Iodide
Format Test Papers
For Use With (Application) pH