Learn More
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 55
Product Code. 11363355
Click to view available options
Diameter (Metric):
25 mm
47 mm
50 mm
100 mm
110 mm
142 mm
Unit Size:
Pack of 100
Pack of 25
Product Code. 11363355
Brand: Cytiva 10410219
Only null left
Add to basket
Add to basket
Consists of pure cellulose without any wetting agents.
  • Spontaneously wetting, very good wet strength
  • Extremely chemically resistant; suitable for aqueous and organic media
  • Hydrophilic
  • Mechanically stable
  • Sterilizable by all methods
TRUSTED_SUSTAINABILITY
Specifications
Material Regenerated Cellulose
Pore Size 0.5 μm
Hydrophobicity Hydrophilic
Thickness (Metric) 75 μm
Diameter (Metric) 100 mm