Learn More
Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD
Product Code. p-4517172
Click to view available options
Pore Size:
0.45 μm
5 μm
10 μm
20 μm
Inlet Style:
With 1/2 in. hose barb (HB) inlet/outlet
With Female NPT inlet/outlet
Unit Size:
Pack of 5
Product Code. 11339104
Brand: Cytiva 2810T
Offers high filtration efficiency and excellent filtrate purity
  • Allows usage with broad range of solutions, pH, and temperature
  • High flow and high retention capacity
  • Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
TRUSTED_SUSTAINABILITY
Specifications
Description Polycap HD 150 Capsule Filter, 0.45 μm, with FNPT inlet and outlet (5 pcs)
Pore Size 0.45 μm
For Use With (Application) Polycap HD 150
Housing Material Polypropylene
Inlet Style With Female NPT inlet/outlet
Max. Pressure 4.1 bar (60 psi)
Quantity 5/Pk.