Learn More
Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD
Product Code. 11359104
Click to view available options
Pore Size:
0.45 μm
5 μm
10 μm
20 μm
Inlet Style:
With 1/2 in. hose barb (HB) inlet/outlet
With Female NPT inlet/outlet
Unit Size:
Pack of 5
Product Code. 11359104
Brand: Cytiva 2810
Offers high filtration efficiency and excellent filtrate purity
  • Allows usage with broad range of solutions, pH, and temperature
  • High flow and high retention capacity
  • Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 0.45 μm
For Use With (Application) Polycap HD 150
Inlet Style With 1/2 in. hose barb (HB) inlet/outlet
Quantity 5/Pk.