Learn More
Sartorius™ Weight Fork
Brand: Sartorius™ YAW42
228.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Specifications
Prevent any damage to the surface of weights | |
1kg Weight |
Product Content Correction
Product Title
Sartorius™ Weight Fork > For Use with 1kg Weight