missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ WaterPro™ Reverse Osmosis (RO) Systems
Three filters (pre, carbon and RO membrane) combined to deliver high-quality reverse osmosis purified water
€ 8560.00
Specifications
Certifications/Compliance | CE, ETL |
---|---|
Capacity (Metric) | 17 L |
Feedwater Source | Tap |
Flow Rate | 1.1 L/min. |
Product Water Type | Type 3 to 4 |
Alert:
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Description
- Ideal for laboratory applications such as reagent preparation and glassware rinsing
- Obtain purified rinse water by connecting to laboratory glassware washers, such as SteamScrubber™ or FlaskScrubber™
- Timed dispense from 1 to 99.9 min. for unattended operation
- Temperature compensated conductivity LED monitor displays water quality in microsiemens from 1 to 999
- Dispense manually from valve or with dispensing gun (sold separately) at rate of 8.7L/min. (tank full) or 1L/min. (tank empty)
- Storage tank air vent filter removes bacteria, maintaining water quality
- Automatic flush valve extends RO membrane life by automatically flushing the RO membrane and tank for three minutes after each 12 hour period of inactivity
- Internal pressure gauge monitors and displays RO-purified water pressure from 0 to 160psi
- Durable Type 304 stainless steel front panel; powder-coated steel cabinet
- Large capacity white polypropylene bowls with threaded heads
- Integral 17L polyethylene storage tank
- Quiet 1/4 hp motor/rotary vane pump
- Wall mountable
Certifications
ETL listed.
Specifications
CE, ETL | |
Tap | |
Type 3 to 4 | |
LED | |
Carbon filtration, reverse osmosis | |
78 cm | |
78 cm | |
19.3 cm | |
40.8 kg |
17 L | |
1.1 L/min. | |
Reverse Osmosis System | |
0 to 160 psi | |
31.1 in. | |
30.8 in. | |
7.6 in. | |
90 lb. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title