Learn More
Water, Reagent (Deionized water), ACS, Thermo Scientific Chemicals Product Code.: 11378078
Water, Reagent (Deionized water), ACS, Thermo Scientific Chemicals
Product Code. 11378078
1 L
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Product Code. 11378078
Brand: Thermo Scientific Alfa Aesar 036645.K2
Request Bulk or Custom Format
Used in conductometric titration, Synthesis of nanomaterials. Mostly it is used as solvent in industries and in all research fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
Specifications
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Solubility Information Miscible in methanol,acetone,Ethanol.
Formula Weight 18.02
Grade ACS
Odor Odorless
Density 1
Quantity 1 L
Refractive Index 1.333
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP25L038 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number036645.M1,036645.K7,036645.K2
Lot NumberW25K017 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number036645.M1,036645.K7,036645.K2
Lot NumberW10K049 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number036645.M1,036645.K7,036645.K2
Lot NumberU06K074 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number036645.M1,036645.K7,036645.K2
Lot NumberU06K072 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number036645.M1,036645.K7,036645.K2
RUO – Research Use Only
Product Content Correction
Product Title
Water, Reagent (Deionized water), ACS, Thermo Scientific Chemicals >